Assay Details
Gene Symbol: REXO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163963.1 | NP_001157435.1 | 15 - 16 | 1722 | 82 | |
XM_023655005.1 | XP_023510773.1 | 1973 | 82 | |||
XM_023655006.1 | XP_023510774.1 | 1977 | 82 | |||
XM_023655007.1 | XP_023510775.1 | 1976 | 82 | |||
GenBank mRNA | AY705653.1 | - | 2 - 3 | 117 | 82 | |
JL624817.1 | - | 4 - 5 | 773 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
