Assay Details
Gene Symbol: CASP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163961.1 | NP_001157433.1 | 5 - 6 | 615 | 91 | |
XM_023630401.1 | XP_023486169.1 | 726 | 91 | |||
GenBank mRNA | DQ174688.1 | - | 2 - 3 | 290 | 91 | |
DQ174689.1 | - | 2 - 3 | 290 | 91 | ||
DQ174690.1 | - | 2 - 3 | 290 | 91 | ||
JL627400.1 | - | 6 - 7 | 639 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
