Assay Details
Gene Symbol: RPL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195513.1 | NP_001182442.1 | 3 - 4 | 244 | 57 | |
GenBank mRNA | CD467943.1 | - | 3 - 4 | 244 | 57 | |
DQ403043.1 | - | 1 - 2 | 108 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
