Assay Details
Gene Symbol: ALOX5AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163965.1 | NP_001157437.1 | 1 - 2 | 130 | 66 | |
XM_005601073.3 | XP_005601130.1 | 190 | 66 | |||
XM_005601074.3 | XP_005601131.1 | 204 | 66 | |||
XM_005601075.3 | XP_005601132.1 | 161 | 66 | |||
XM_023621300.1 | XP_023477068.1 | 207 | 66 | |||
XM_023621301.1 | XP_023477069.1 | 177 | 66 | |||
XM_023621302.1 | XP_023477070.1 | 172 | 66 | |||
GenBank mRNA | M96552.1 | - | 1 - 2 | 72 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
