Assay Details
Gene Symbol: RGS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144133.1 | NP_001137605.1 | 2 - 3 | 218 | 72 | |
GenBank mRNA | AB278562.1 | - | 2 - 3 | 218 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
