Assay Details
Gene Symbol: LDHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144880.1 | NP_001138352.1 | 2 - 3 | 233 | 66 | |
XM_023644137.1 | XP_023499905.1 | 230 | 66 | |||
GenBank mRNA | AB472335.1 | - | 2 - 3 | 233 | 66 | |
AB477533.1 | - | 2 - 3 | 233 | 66 | ||
GU808337.1 | - | 2 - 3 | 233 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
