Assay Details
Gene Symbol: GNRHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081836.1 | NP_001075305.1 | 3 | 947 | 97 | |
GenBank mRNA | AF018072.3 | - | 3 | 939 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
