Assay Details
Gene Symbol: OAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082489.1 | NP_001075958.1 | 1 | 151 | 107 | |
GenBank mRNA | AY321355.1 | - | 1 | 151 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
