Assay Details
Gene Symbol: JAZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023639427.1 | XP_023495195.1 | 667 | 79 | ||
XM_023639428.1 | XP_023495196.1 | 946 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
