Assay Details
Gene Symbol: PTHLH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163981.1 | NP_001157453.1 | 2 - 3 | 317 | 68 | |
XM_014740461.2 | XP_014595947.1 | 465 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001163981.1, XM_014740461.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
