Assay Details
Gene Symbol: TNFSF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205160.1 | NP_001192089.1 | 3 - 4 | 382 | 68 | |
XM_005597024.3 | XP_005597081.1 | 402 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
