Assay Details
Gene Symbol: UBE2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242449.2 | NP_001229378.1 | 6 - 7 | 629 | 75 | |
XM_005602131.2 | XP_005602188.1 | 412 | 75 | |||
XM_005602132.3 | XP_005602189.1 | 386 | 75 | |||
XM_005602133.3 | XP_005602190.1 | 335 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
