Assay Details
Gene Symbol: ARF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242541.2 | NP_001229470.1 | 3 - 4 | 399 | 79 | |
XM_005597022.3 | XP_005597079.1 | 504 | 79 | |||
GenBank mRNA | JL621768.1 | - | 3 - 4 | 285 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
