Assay Details
Gene Symbol: SH3BP5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242551.1 | NP_001229480.1 | 2 - 3 | 251 | 77 | |
XM_005604190.3 | XP_005604247.1 | 829 | 77 | |||
XM_014734839.2 | XP_014590325.1 | 525 | 77 | |||
XM_014734840.2 | XP_014590326.1 | 370 | 77 | |||
XM_023625241.1 | XP_023481009.1 | 261 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023625241.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
