Assay Details
Gene Symbol: SH3BP5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242551.1 | NP_001229480.1 | 5 - 6 | 705 | 65 | |
XM_005604190.3 | XP_005604247.1 | 1283 | 65 | |||
XM_014734839.2 | XP_014590325.1 | 979 | 65 | |||
XM_014734840.2 | XP_014590326.1 | 824 | 65 | |||
XM_023625240.1 | XP_023481008.1 | 1091 | 65 | |||
XM_023625241.1 | XP_023481009.1 | 715 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
