Assay Details
Gene Symbol: ARHGEF39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195761.1 | NP_001182690.1 | 2 - 3 | 238 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ARHGEF39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195761.1 | NP_001182690.1 | 2 - 3 | 238 | 62 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |