Assay Details
Gene Symbol: SOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082517.2 | NP_001075986.1 | 2 - 3 | 296 | 96 | |
GenBank mRNA | AB001693.1 | - | 2 - 3 | 296 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
