Assay Details
Gene Symbol: SLC25A38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501962.4 | XP_001502012.1 | 631 | 88 | ||
XM_023620706.1 | XP_023476474.1 | 721 | 88 | |||
XM_023620707.1 | XP_023476475.1 | 640 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
