Assay Details
Gene Symbol: CNOT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001917441.5 | XP_001917476.3 | 910 | 102 | ||
XM_023617361.1 | XP_023473129.1 | 836 | 102 | |||
XM_023617362.1 | XP_023473130.1 | 815 | 102 | |||
XM_023617363.1 | XP_023473131.1 | 703 | 102 | |||
XM_023617364.1 | XP_023473132.1 | 668 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
