Assay Details
Gene Symbol: SOX30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005614964.3 | XP_005615021.3 | 1537 | 66 | ||
XM_023617321.1 | XP_023473089.1 | 1547 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
