Assay Details
Gene Symbol: DTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005611346.3 | XP_005611403.2 | 1260 | 72 | ||
XM_005611348.3 | XP_005611405.2 | 1271 | 72 | |||
XM_014740943.2 | XP_014596429.1 | 1705 | 72 | |||
XM_014740944.2 | XP_014596430.1 | 1716 | 72 | |||
XM_023643882.1 | XP_023499650.1 | 1648 | 72 | |||
XM_023643883.1 | XP_023499651.1 | 1452 | 72 | |||
XM_023643884.1 | XP_023499652.1 | 1313 | 72 | |||
XM_023643885.1 | XP_023499653.1 | 1331 | 72 | |||
XM_023643886.1 | XP_023499654.1 | 1327 | 72 | |||
XM_023643887.1 | XP_023499655.1 | 1324 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
