Assay Details
Gene Symbol: RBPMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001915723.5 | XP_001915758.1 | 516 | 75 | ||
XM_003364229.4 | XP_003364277.1 | 516 | 75 | |||
XM_005606347.3 | XP_005606404.1 | 516 | 75 | |||
XM_014736587.2 | XP_014592073.1 | 516 | 75 | |||
XM_014736588.2 | XP_014592074.1 | 516 | 75 | |||
XM_014736589.2 | XP_014592075.1 | 477 | 75 | |||
XM_014736590.2 | XP_014592076.1 | 625 | 75 | |||
XM_023630819.1 | XP_023486587.1 | 516 | 75 | |||
XM_023630821.1 | XP_023486589.1 | 516 | 75 | |||
XM_023630822.1 | XP_023486590.1 | 586 | 75 | |||
XM_023630825.1 | XP_023486593.1 | 516 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
