Assay Details
Gene Symbol: SNRPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253840.1 | NP_001240769.1 | 6 - 7 | 590 | 78 | |
XM_005604412.3 | XP_005604469.1 | 703 | 78 | |||
XM_005604413.3 | XP_005604470.1 | 984 | 78 | |||
XM_005604414.3 | XP_005604471.1 | 684 | 78 | |||
XM_014734997.2 | XP_014590483.1 | 583 | 78 | |||
XM_023625917.1 | XP_023481685.1 | 861 | 78 | |||
GenBank mRNA | HQ889944.1 | - | 6 - 7 | 590 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
