Assay Details
Gene Symbol: ENO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267603.3 | NP_001254532.1 | 5 - 6 | 398 | 101 | |
XM_023626513.1 | XP_023482281.1 | 465 | 101 | |||
GenBank mRNA | JQ713943.1 | - | 5 - 6 | 401 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
