Assay Details
Gene Symbol: MRPL45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286864.1 | NP_001273793.1 | 6 - 7 | 670 | 67 | |
GenBank mRNA | HQ890268.1 | - | 6 - 7 | 673 | 67 | |
JL615914.1 | - | 6 - 7 | 670 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
