Assay Details
Gene Symbol: METTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301208.1 | NP_001288137.1 | 1 - 2 | 97 | 56 | |
XM_014740756.2 | XP_014596242.1 | 262 | 56 | |||
XM_023642189.1 | XP_023497957.1 | 263 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
