Assay Details
Gene Symbol: ZMPSTE24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301234.1 | NP_001288163.1 | 3 - 4 | 355 | 63 | |
XM_005607173.3 | XP_005607230.1 | 427 | 63 | |||
XM_014737160.2 | XP_014592646.2 | 575 | 63 | |||
GenBank mRNA | JL639173.1 | - | 2 - 3 | 243 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
