Assay Details
Gene Symbol: RAP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301245.1 | NP_001288174.1 | 2 - 3 | 162 | 137 | |
GenBank mRNA | JL618129.1 | - | 2 - 3 | 162 | 137 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001301245.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
