Assay Details
Gene Symbol: EIF3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301291.1 | NP_001288220.1 | 11 - 12 | 1197 | 77 | |
GenBank mRNA | CX601408.1 | - | 6 - 7 | 664 | 77 | |
JL616236.1 | - | 5 - 6 | 499 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
