Assay Details
Gene Symbol: GPR171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308940.1 | NP_001295869.1 | 1 - 2 | 117 | 103 | |
GenBank mRNA | CD466857.1 | - | 1 - 2 | 116 | 103 | |
JL616296.2 | - | 1 - 2 | 122 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001308940.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
