Assay Details
Gene Symbol: ARL6IP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308942.1 | NP_001295871.1 | 1 - 2 | 290 | 78 | |
GenBank mRNA | JG392607.1 | - | 1 - 2 | 290 | 78 | |
JL618168.1 | - | 1 - 2 | 311 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
