Assay Details
Gene Symbol: RHOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308946.1 | NP_001295875.1 | 2 - 3 | 237 | 71 | |
XM_023619803.1 | XP_023475571.1 | 508 | 71 | |||
XM_023619804.1 | XP_023475572.1 | 254 | 71 | |||
XM_023619805.1 | XP_023475573.1 | 290 | 71 | |||
GenBank mRNA | JL616266.1 | - | 3 - 4 | 336 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
