Assay Details
Gene Symbol: RAB18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308960.1 | NP_001295889.1 | 4 - 5 | 272 | 72 | |
XM_023631885.1 | XP_023487653.1 | 432 | 72 | |||
GenBank mRNA | JL616370.2 | - | 4 - 5 | 272 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
