Assay Details
Gene Symbol: CCND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309189.1 | NP_001296118.1 | 4 - 5 | 1008 | 54 | |
GenBank mRNA | CD467520.1 | - | 1 - 2 | 109 | 54 | |
CX605335.1 | - | 4 - 5 | 643 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
