Assay Details
Gene Symbol: STK17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309194.1 | NP_001296123.1 | 5 - 6 | 680 | 79 | |
XM_023621972.1 | XP_023477740.1 | 777 | 79 | |||
GenBank mRNA | JL616416.2 | - | 5 - 6 | 676 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
