Assay Details
Gene Symbol: AIMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309207.1 | NP_001296136.1 | 2 - 3 | 253 | 73 | |
XM_023627008.1 | XP_023482776.1 | 392 | 73 | |||
GenBank mRNA | JL615910.1 | - | 3 - 4 | 291 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
