Assay Details
Gene Symbol: UBE2E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309219.1 | NP_001296148.1 | 3 - 4 | 403 | 116 | |
XM_005601618.3 | XP_005601675.1 | 556 | 116 | |||
XM_005601619.3 | XP_005601676.1 | 528 | 116 | |||
XM_023621981.1 | XP_023477749.1 | 677 | 116 | |||
GenBank mRNA | HQ889972.1 | - | 3 - 4 | 333 | 116 | |
JL616154.1 | - | 3 - 4 | 403 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
