Assay Details
Gene Symbol: PPIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309220.1 | NP_001296149.1 | 6 - 7 | 469 | 85 | |
XM_023621982.1 | XP_023477750.1 | 531 | 85 | |||
GenBank mRNA | DN506060.1 | - | 6 - 7 | 468 | 85 | |
JL615875.1 | - | 5 - 6 | 438 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
