Assay Details
Gene Symbol: SEC22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309225.1 | NP_001296154.1 | 3 - 4 | 386 | 78 | |
XM_023623146.1 | XP_023478914.1 | 623 | 78 | |||
GenBank mRNA | JL615908.2 | - | 3 - 4 | 396 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
