Assay Details
Gene Symbol: ECHDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309234.1 | NP_001296163.1 | 5 - 6 | 641 | 79 | |
XM_023649403.1 | XP_023505171.1 | 757 | 79 | |||
XM_023649404.1 | XP_023505172.1 | 740 | 79 | |||
XM_023649405.1 | XP_023505173.1 | 620 | 79 | |||
XM_023649406.1 | XP_023505174.1 | 598 | 79 | |||
GenBank mRNA | HQ890193.1 | - | 5 - 6 | 888 | 79 | |
JL616276.1 | - | 5 - 6 | 650 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
