Assay Details
Gene Symbol: ARPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309270.1 | NP_001296199.1 | 5 - 6 | 394 | 68 | |
GenBank mRNA | JL616131.1 | - | 5 - 6 | 394 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
