Assay Details
Gene Symbol: BET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309271.1 | NP_001296200.1 | 1 - 2 | 151 | 100 | |
GenBank mRNA | CD470418.1 | - | 1 - 2 | 151 | 100 | |
JL616348.2 | - | 1 - 2 | 126 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001309271.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
