Assay Details
Gene Symbol: BET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309271.1 | NP_001296200.1 | 2 - 3 | 275 | 89 | |
GenBank mRNA | CD470418.1 | - | 2 - 3 | 275 | 89 | |
JL616348.2 | - | 2 - 3 | 250 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
