Assay Details
Gene Symbol: ACAA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309272.1 | NP_001296201.1 | 6 - 7 | 782 | 60 | |
GenBank mRNA | JL616308.1 | - | 6 - 7 | 784 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
