Assay Details
Gene Symbol: HAUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309277.1 | NP_001296206.1 | 5 - 6 | 660 | 91 | |
XM_005612873.3 | XP_005612930.1 | 963 | 91 | |||
GenBank mRNA | JL615909.2 | - | 5 - 6 | 634 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
