Assay Details
Gene Symbol: BLVRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309290.1 | NP_001296219.1 | 3 - 4 | 275 | 76 | |
XM_005609106.3 | XP_005609163.1 | 311 | 76 | |||
XM_005609107.3 | XP_005609164.1 | 333 | 76 | |||
XM_014738889.2 | XP_014594375.2 | 879 | 76 | |||
XM_023638772.1 | XP_023494540.1 | 743 | 76 | |||
XM_023638773.1 | XP_023494541.1 | 713 | 76 | |||
XM_023638774.1 | XP_023494542.1 | 325 | 76 | |||
GenBank mRNA | JL616290.1 | - | 3 - 4 | 301 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
