Assay Details
Gene Symbol: NDUFA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309295.1 | NP_001296224.1 | 2 - 3 | 174 | 81 | |
GenBank mRNA | JL638426.1 | - | 2 - 3 | 76 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
