Assay Details
Gene Symbol: PRPF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309307.1 | NP_001296236.1 | 3 - 4 | 251 | 78 | |
XM_023631887.1 | XP_023487655.1 | 398 | 78 | |||
GenBank mRNA | JL618178.1 | - | 3 - 4 | 251 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
