Assay Details
Gene Symbol: GTF2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309309.1 | NP_001296238.1 | 2 - 3 | 193 | 75 | |
GenBank mRNA | JL641402.1 | - | 2 - 3 | 193 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
