Assay Details
Gene Symbol: DCTN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309312.1 | NP_001296241.1 | 3 - 4 | 201 | 74 | |
XM_023630405.1 | XP_023486173.1 | 298 | 74 | |||
GenBank mRNA | JL616465.2 | - | 3 - 4 | 201 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
